Order Entry
Czech Republic
ContactUsLinkComponent
2-(2-Ethoxyethoxy)ethyl acetate, Sigma-Aldrich®
2-(2-Ethoxyethoxy)ethyl acetate, Sigma-Aldrich®
Catalog # SIAL32240-1L
Supplier:  Merck
CAS Number:  
2-(2-Ethoxyethoxy)ethyl acetate, Sigma-Aldrich®
Catalog # SIAL32240-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :176.21 g/mol
Boiling Pt:210 °C (1013 hPa)
Melting Pt:–25 °C
Density:1.0114 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:203-940-1
CAS Number:112-15-2
Formula:C₈H₁₆O₄
MDL Number:MFCD00041928