Order Entry
Czech Republic
ContactUsLinkComponent
Palladium(II) acetate, Sigma-Aldrich®
Palladium(II) acetate, Sigma-Aldrich®
Catalog # SIAL720070-1G
Supplier:  Merck
CAS Number:  
Palladium(II) acetate, Sigma-Aldrich®
Catalog # SIAL720070-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :224.51 g/mol
Melting Pt:205 °C
EINECS:222-164-4
CAS Number:3375-31-3
Formula:C₄H₆O₄Pd
MDL Number:MFCD00012453