Cerium triacetate hydrate, Sigma-Aldrich®
Catalog # SIAL529559-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:335.266 g/mol
:M-Clarity™ quality level = MQ100
EINECS:208-654-0
CAS Number:206996-60-3
:537-00-8
Formula:C₆H₉CeO₆·H₂O
MDL Number:MFCD00150533