Acetonitrile
Glass bottle, 50 mL
Supplier: Thermo Fisher Scientific
Acetonitrile
Catalog # HYPETS-20062
Supplier: Thermo Fisher Scientific
CAS Number: 75-05-8
Jump to:
Product Details & Documents
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- CAS Number:75-05-8
- Formula:H₃CCN
- MDL Number:MFCD00001878
- Molecular Weight:41.05 g/mol
- Merck Index:13,00071
- Boiling Pt:81.6 °C (1013 hPa)
- Melting Pt:–45.7 °C
- Density:0.782 g/cm³ (20 °C)
- Flash Pt:2 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-835-2
- UN:1648
- ADR:3,II
- REACH:01-2119471307-38
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Cat. No.:HYPETS-20062
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health



