Dexamethasone, Sigma-Aldrich®
Catalog # SIAL31375-500MG
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:500 mg
- Sekvence:2725
Specifications
About this item
:
Melting Pt:268…271 °C
:M-Clarity™ quality level = MQ100
EINECS:200-003-9
CAS Number:50-02-2
Formula:C₂₂H₂₉FO₅
MDL Number:MFCD00064136