5-Chloro-3-methyl-1-phenyl-1H-pyrazole ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1131-17-5
:
Formula:C₁₀H₉ClN₂
MDL Number:MFCD00067853
:192.65 g/mol
Boiling Pt:114…115 °C (4 mmHg)
Storage Temperature:Ambient
Related Information
Frequently Bought Together