4-Methoxybenzamide, Sigma-Aldrich®
Catalog # SIALM10069-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:151.16 g/mol
Boiling Pt:295 °C (1013 hPa)
Melting Pt:130 °C
:M-Clarity™ quality level = MQ100
EINECS:222-319-6
CAS Number:3424-93-9
Formula:C₈H₉NO₂
MDL Number:MFCD00007995