
4-Acetamidobenzenesulphonyl chloride 99%
:121-60-8
:
:C₈H₈ClNO₃S
:MFCD00007442
:233.68 g/mol
:416 °C (1013 hPa)
:145…148 °C (dec.)
:>112 °C
:Ambient
:204-485-1
:3261
:8,II
:GHS05
- Appearance:White to cream to light beige Crystalline or granular powder or crystals
- Infrared spectrum:Conforms
- Melting point:144 to 148 °C (Dec)
- Titration Argentometric:≥98.5 %
Frequently Bought Together