
4-(2-Hydroxyethyl)pyridine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5344-27-4
:
Formula:C₇H₉NO
MDL Number:MFCD00129038
:123.15 g/mol
Boiling Pt:121…122 °C (2 mmHg)
Melting Pt:8…10 °C
Density:1.093
Flash Pt:92 °C (197 °F)
EINECS:226-286-9
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together