Ethyl-3-phenylpropionate ≥98%
A13055.18
:
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
: Ethyl 3-Phenylpropionate, Ethyl hydrocinnamate, 3-Phenylpropionic acid ethyl ester, Ethyl dihydrocinnamate, Hydrocinnamic acid ethyl ester
:2021-28-5
:
:C₁₁H₁₄O₂
:MFCD00009206
:178.23 g/mol
:118 °C (3 mmHg)
:1.016
:107 °C (224 °F)
:Ambient
:217-966-6