
3-Fluoro-4-nitrobenzenesulphonyl chloride ≥98%
Catalog # H31979.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:239.61 g/mol
Melting Pt:44…48 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
CAS Number:86156-93-6
Formula:C₆H₃ClFNO₄S
MDL Number:MFCD03412240