Acetonitrile ≥99.9% (by GC), CHROMASOLV™, gradient grade suitable for PAH analysis, Riedel-de Haën™
Katalogové číslo 00683-2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay (GC):Min. 99.9 %
- Density (D 20/4):0.765 - 0.794
- Refractive index (n 20/D):1.343 - 1.345
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.02 %
- Free acid (as CH₃COOH):Max. 0.005 %
- Free alkali (as NH₃):Max. 0.0005 %
- Suitability test (HPLC):complying
- Absorbance at 230 nm:Max. 0.005
- Absorbance at 215 nm:Max. 0.01
- Absorbance at 200 nm:Max. 0.04
- Fluorescent substances:complying
- Refractive Index:n20/D 1.344 (lit.)
Specification Test Results
About this item
:
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:82 °C (1.013 hPa)
Melting Pt:–46 °C
Density:0.7857 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS08
:GHS07
:GHS02
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878