

cis-1,2-Cyclohexanedicarboxylic anhydride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13149-00-3
:
Formula:C₈H₁₀O₃
MDL Number:MFCD00005926
Boiling Pt:296 °C (760 mmHg)
Melting Pt:29…32 °C
Flash Pt:143 °C
EINECS:236-086-3
:GHS08
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Low melting crystalline solid
- Infrared spectrum:Conforms
- Melting point:29 to 32 °C
- GC:≥98.5 %
- Color of molten substance:≤50 APHA
Specification Test Results
Frequently Bought Together