3,4-Dibutoxy-3-cyclobutene-1,2-dione, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2892-62-8
:
Formula:C₁₂H₁₈O₄
MDL Number:MFCD00037150
:226.27 g/mol
Boiling Pt:121 °C (1013 hPa)
Density:0.96 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Frequently Bought Together