
3,4-Dibutoxy-3-cyclobutene-1,2-dione 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2892-62-8
:
Formula:C₁₂H₁₈O₄
MDL Number:MFCD00037150
:226.27 g/mol
Boiling Pt:121…122 °C (.2 mmHg)
Density:1.05 g/cm³
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4930 to 1.4950 (20°C, 589 nm)
- GC:≥96.0 %
- NMR (13C-NMR):Consistent with structure
- NMR (31P-NMR):Consistent with structure
- Elemental Analysis:76.56 to 82.94 % C
Specification Test Results
Frequently Bought Together