3-(Methylthio)-1-propanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:505-10-2
:
Formula:C₄H₁₀OS
MDL Number:MFCD00036560
:106.19 g/mol
Boiling Pt:195 °C (1013 hPa)
Density:1.03 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:208-004-6
Frequently Bought Together