Order Entry
Czech Republic
ContactUsLinkComponent
Naphthol AS-D chloroacetate
Naphthol AS-D chloroacetate
Catalog # A4454.0001
Supplier:  PanReac AppliChem
CAS Number:  
Naphthol AS-D chloroacetate
Catalog # A4454.0001
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :353.8 g/mol
EINECS:252-463-5
CAS Number:35245-26-2
Formula:C₂₀H₁₆ClNO₃