
2-Hydroxybenzimidazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:615-16-7
:
Formula:C₇H₆N₂O
MDL Number:MFCD00127894
:134.14 g/mol
Melting Pt:314…316 °C
Density:1,42 g/cm³ (20 °C)
EINECS:210-412-4
UN:2811
:GHS07
Specification Test Results
- Appearance:Off-white to light brown Crystalline powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥96.0 %
- GC:≥94 %
Specification Test Results
Frequently Bought Together