2-Hydroxybenzimidazole
Catalog # PROOMM0474.03
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:134.14 g/mol
Melting Pt:320…322 °C
Density:1,42 g/cm³ (20 °C)
EINECS:210-412-4
CAS Number:615-16-7
Formula:C₇H₆N₂O
MDL Number:MFCD00127894