2-Fluoro-4-(trifluoromethyl)pyridine, Sigma-Aldrich®
Catalog # SIAL716294-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:165.09 g/mol
CAS Number:118078-66-3
Formula:C₆H₃F₄N
MDL Number:MFCD09879262