
2-Ethylhexane-1,3-diol (mixture of isomers) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:94-96-2
:
Formula:C₈H₁₈O₂
MDL Number:MFCD00004578
:146.23 g/mol
Boiling Pt:244 °C (1013 hPa)
Melting Pt:–40 °C
Density:0.9422 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:202-377-9
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:42 g/l (20 °C)
- Melting Point:-40 °C
- Molar Mass:146.23 g/mol
- Boiling Point:241 - 249 °C (1013 hPa)
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:136 °C
- Refractive Index:1.4530 (22 °C, 589 nm)
- Density:0.94 g/cm3 (20 °C)
- Ignition Point:340 °C DIN 51794
Specification Test Results
Frequently Bought Together