Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Ethylhexane-1,3-diol (mixture of isomers)
2-Ethylhexane-1,3-diol (mixture of isomers)
2-Ethylhexane-1,3-diol (mixture of isomers)
EHERC13340000
:
EHRENSTORFER
:
2-Ethylhexane-1,3-diol (mixture of isomers)
EHERC13340000
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
146.23 g/mol
:
244 °C (1013 hPa)
:
–40 °C
:
0,9422 g/cm³ (20 °C)
:
202-377-9
:
94-96-2
:
C₈H₁₈O₂
:
MFCD00004578