Order Entry
Czech Republic
ContactUsLinkComponent
2-Acetyl-1-naphthol 99%
Irritant
2-Acetyl-1-naphthol 99%
Certificates
undefined
Irritant
2-Acetyl-1-naphthol 99%

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    97 to 100 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together