Order Entry
Czech Republic
ContactUsLinkComponent
2,5-Dibromo-4-picoline 98%
Irritant
2,5-Dibromo-4-picoline 98%
Certificates
undefined
Irritant
2,5-Dibromo-4-picoline 98%

Specification Test Results

  • Appearance:
    Light yellow to orange Low melting solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    37 to 41 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together