


2,3-Dichloro-5-(trifluoromethyl)pyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69045-84-7
:
Formula:C₆H₂Cl₂F₃N
MDL Number:MFCD00042243
:215.99 g/mol
Boiling Pt:176 °C (760 mmHg)
Melting Pt:8…9 °C
Density:1.549 g/cm³
Flash Pt:79 °C
Storage Temperature:Ambient
EINECS:410-340-5
UN:3082
ADR:9,III
:GHS05
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4745 to 1.4765 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together