

2,3,6-Trifluoro-4-(trifluoromethyl)pyridine ≥97%
Catalog # L20053.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:201.07 g/mol
Boiling Pt:104…106 °C
Density:1.579
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:84940-46-5
Formula:C₆HF₆N
MDL Number:MFCD03840203