Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Isopropylimidazole
2-Isopropylimidazole, Sigma-Aldrich®
2-Isopropylimidazole, Sigma-Aldrich®
2-Isopropylimidazole, Sigma-Aldrich®
SIAL373990-100G
:
Merck
:
2-Isopropylimidazole, Sigma-Aldrich®
SIAL373990-100G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
110.16 g/mol
:
M-Clarity™ quality level = MQ100
:
253-286-6
:
36947-68-9
:
C₆H₁₀N₂
:
MFCD00014486