Pesticide reference standard, 2,4,6-Tribromophenol 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL17666000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:118-79-6
- Description:2,4,6-Tribromophenol 10 µg/ml in methanol
- Formula:C₆H₃Br₃O
- Description:2,4,6-Tribromophenol