Order Entry
Czech Republic
ContactUsLinkComponent
Pesticide reference standard, 2,3,4,5-Tetrachlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, 2,3,4,5-Tetrachlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17374500CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 2,3,4,5-Tetrachlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17374500CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    4901-51-3
  • Description:
    2,3,4,5-Tetrachlorophenol 10 µg/ml in cyclohexane
  • Formula:
    C₆H₂Cl₄O
  • Description:
    2,3,4,5-Tetrachlorophenol

Specifications

About this item