3'-Fluoro-4'-methoxyacetophenone, Sigma-Aldrich®
Catalog # SIAL361836-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:168.17
Boiling Pt:147…148 °C (20 mmHg)
Melting Pt:92 °C
:M-Clarity™ quality level = MQ100
EINECS:207-253-8
CAS Number:455-91-4
Formula:C₉H₉FO₂
MDL Number:MFCD00026219