
3'-Fluoro-4'-methoxyacetophenone ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:455-91-4
:
Formula:C₉H₉FO₂
MDL Number:MFCD00026219
:168.17
Boiling Pt:147…148 °C (20 mmHg)
Melting Pt:90…94 °C
Storage Temperature:Ambient
EINECS:207-253-8
:GHS07
Related Information
Frequently Bought Together