
2'-Hydroxy-5'-methylacetophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1450-72-2
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00002380
:150.18 g/mol
Boiling Pt:112…114 °C (12 mmHg)
Melting Pt:45…48 °C
Density:1.079
Flash Pt:>110 °C (230 °F)
EINECS:215-915-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together