
2'-Hydroxy-5'-methyl-3'-nitroacetophenone, Sigma-Aldrich®
Catalog # SIAL415766-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:195.17 g/mol
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:66108-30-3
Formula:C₉H₉NO₄
MDL Number:MFCD00192216