
3,5-Difluoro-4-hydroxybenzonitrile ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2967-54-6
:
Formula:C₇H₃F₂NO
MDL Number:MFCD09033181
:155.1 g/mol
Melting Pt:200…208 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together