3,4-Dimethoxybenzonitrile, Sigma-Aldrich®
SIALD132306-25G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
:
:163.18 g/mol
:131…133 °C (0.3 mmHg)
:71…72 °C
:6.1,III
:M-Clarity™ quality level = MQ100
:217-969-2
:3439
:2024-83-1
:C₉H₉NO₂
:MFCD00001802