4'-Chloro-3'-nitroacetophenone, Sigma-Aldrich®
Catalog # SIAL193968-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:199.59
:M-Clarity™ quality level = MQ100
EINECS:226-769-4
CAS Number:5465-65-6
Formula:C₈H₆ClNO₃
MDL Number:MFCD00007083