2-(4-Chlorophenylthio)ethanol, Sigma-Aldrich®
Catalog # 8.14889.0005
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 ml
- Seq:2210
Specifications
Specification Test Results
- Molar Mass:188.68 g/mol
- Boiling Point:140 °C (1013 hPa)
- Density:1.278 g/cm3 (20 °C)
- Hill Formula:C8H9ClOS
Specification Test Results
About this item
:
:188.68 g/mol
Boiling Pt:305 °C (1013 hPa)
Melting Pt:41 °C
Density:1.07 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-657-7
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ200
CAS Number:13457-98-2
Formula:C₈H₉ClOS
MDL Number:MFCD00014037