Amantadine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:665-66-7
:
Formula:C₁₀H₁₈ClN
MDL Number:MFCD00074723
:187.71 g/mol
Melting Pt:360 °C
:M-Clarity™ quality level = MQ100
EINECS:211-560-2
Frequently Bought Together