

2,6-Dimethyl-4-fluoroaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:392-70-1
:
Formula:C₈H₁₀FN
MDL Number:MFCD03792669
:139.17 g/mol
:GHS06
:GHS07
Storage Temperature:Ambient
Frequently Bought Together