Order Entry
Czech Republic
ContactUsLinkComponent
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Flammable
Irritant
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Solubility:
    (20 °C) 
  • Melting Point:
    -45.7 °C
  • Molar Mass:
    41.05 g/mol
  • Boiling Point:
    81.6 °C (1013 hPa)
  • Vapor Pressure:
    97 hPa (20 °C)
  • Flash Point:
    2 °C
  • Refractive Index:
    1.34
  • Explosion Limit:
    3.0 - 17 %(V)
  • Density:
    0.786 g/cm³ (20 °C)
  • Ignition Point:
    524 °C
  • Synonyms:
    ACN, Methyl cyanide, Ethyl nitrile, Cyanomethane
  • Hill Formula:
    C₂H₃N
  • Chemical Formula:
    CH₃CN

Specification Test Results

Frequently Bought Together