1,4-Diaminobutane dihydrochloride, Sigma-Aldrich®
Catalog # SIALP6024-1MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
Density:1.31 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:161.07 g/mol
EINECS:206-375-9
CAS Number:333-93-7
Formula:C₄H₁₄Cl₂N₂
MDL Number:MFCD00012526