1,4-Diaminobutane dihydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:333-93-7
:
Formula:C₄H₁₄Cl₂N₂
MDL Number:MFCD00012526
Density:1.31 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:161.07 g/mol
EINECS:206-375-9
Frequently Bought Together