Order Entry
Czech Republic
ContactUsLinkComponent
3',5'-Dimethoxy-4'-hydroxyacetophenone 97%
Irritant
3',5'-Dimethoxy-4'-hydroxyacetophenone 97%
Certificates
undefined
Irritant
3',5'-Dimethoxy-4'-hydroxyacetophenone 97%

Specification Test Results

  • Appearance:
    Beige to light brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    123 to 128 °C
  • GC:
    ≥96.0 %
  • Titration with NaOH:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together