2,4-Dichloro-1-naphthol, Sigma-Aldrich®
SIAL251976-25G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
:
:213.06 g/mol
:340 °C (1013 hPa)
:104…106 °C
:M-Clarity™ quality level = MQ100
:218-103-6
:2050-76-2
:C₁₀H₆Cl₂O
:MFCD00003931