
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:328-73-4
:
Formula:C₈H₃F₆I
MDL Number:MFCD00040837
:340.01 g/mol
Boiling Pt:62…64 °C (12 mmHg)
Density:1.919
Flash Pt:74 °C (165 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together