
3',5'-Bis(trifluoromethyl)acetophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:30071-93-3
:
Formula:C₁₀H₆F₆O
MDL Number:MFCD00009910
:256.15 g/mol
Boiling Pt:85 °C (8 mmHg)
Density:1.422
Flash Pt:82 °C (179 °F)
EINECS:250-023-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together