3',5'-Bis(trifluoromethyl)acetophenone, Sigma-Aldrich®
Catalog # SIAL263362-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:256.15 g/mol
Boiling Pt:95 °C (15 mmHg)
Density:1.42 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:250-023-7
CAS Number:30071-93-3
Formula:C₁₀H₆F₆O
MDL Number:MFCD00009910