4'-tert-Butylacetophenone, Sigma-Aldrich®
Catalog # SIAL357642-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:176.26
Boiling Pt:71…72 °C (0.09 mmHg)
Melting Pt:18 °C
Density:0.9705 g/cm³ (0 °C)
:M-Clarity™ quality level = MQ100
EINECS:213-399-3
CAS Number:943-27-1
Formula:C₁₂H₁₆O
MDL Number:MFCD00017256