Acetonitrile, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
EINECS:200-835-2
UN:1648
ADR:3,II
:M-Clarity™ quality level = MQ300
Flash Pt:2 °C (closed cup)
REACH:01-2119471307-38
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-45.7 °C
- Molar Mass:41.05 g/mol
- Boiling Point:81.6 °C (1013 hPa)
- Vapor Pressure:97 hPa (20 °C)
- Flash Point:2 °C
- Refractive Index:1.34
- Explosion Limit:3.0 - 17 %(V)
- Density:0.786 g/cm3 (20 °C)
- Ignition Point:524 °C
Specification Test Results
Related Information
Frequently Bought Together