
Tyramine hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60-19-5
:
Formula:C₈H₁₂ClNO
MDL Number:MFCD00012901
:173.64
Boiling Pt:269 °C (760 mmHg)
Melting Pt:271…274 °C
Storage Temperature:Ambient
EINECS:200-462-5
:GHS07
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:98.5 to 101.5 % (on dry substance)
- Solubility:(10 % in water) Clear colorless to light yellow
Specification Test Results
Frequently Bought Together